Learn More
Description
Specifications
Specifications
| Antigen | DEC1 |
| Applications | Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:5000-1:15000, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature (PMID 29715265), Flow Cytometry 2-5 ug/ml, ELISA reported in scientific literature (PMID 31061528), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Chromatin Immunoprecipitation (ChIP), Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 31061528) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | O14503 |
| Gene Alias | basic helix-loop-helix domain containing, class B, 2, basic helix-loop-helix family, member e40, bHLHb2, bHLHe40, Class B basic helix-loop-helix protein 2, class E basic helix-loop-helix protein 40, DEC1HLHB2, differentially expressed in chondrocytes 1, Differentially expressed in chondrocytes protein 1, differentiated embryo chondrocyte expressed gene 1, Enhancer-of-split and hairy-related protein 2, SHARP2, SHARP-2, Stimulated by retinoic acid gene 13 protein, STRA13FLJ99214, Stra14 |
| Gene Symbols | BHLHE40 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
