Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D(+)-Mannose, 99+%

Catalog No. AC150600250
Click to view available options
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Plastic drum

Catalog No. AC150600250

Supplier: Thermo Scientific Chemicals 150600250

Only null left
Add to Cart

CAS: 3458-28-4 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-28-4
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers

Specifications

Melting Point 133.0°C
Color White
Loss on Drying 0.45% max.
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Odor Odorless
Packaging Plastic bottle
Quantity 25 g
Beilstein 31284
Merck Index 155812
Specific Rotation Condition + 14.00 (20.00°C 589nm,c=10,H2O)
Specific Rotation + 14.00
Solubility Information Solubility in water: 2480g/L (17°C)
Formula Weight 180.16
Percent Purity 99+%
Physical Form Crystals or Powder
Chemical Name or Material D(+)-Mannose
Show More Show Less
EINECSNumber 222-392-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.