Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Malic acid, 98%

Catalog No. ac153500250
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

Catalog No. AC153500250

Supplier: Thermo Scientific Chemicals 153500250

Only null left
Add to Cart

CAS: 636-61-3 | C4H6O5 | 134.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-61-3
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.09
MDL Number MFCD00004245
InChI Key BJEPYKJPYRNKOW-UHFFFAOYNA-N
Synonym d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate
PubChem CID 92824
ChEBI CHEBI:30796
IUPAC Name (2R)-2-hydroxybutanedioic acid
SMILES OC(CC(O)=O)C(O)=O

Specifications

Melting Point 99.0°C to 104.0°C
Color White
Assay Percent Range 98%
Linear Formula HO2CCH2CH(OH)CO2H
Beilstein 03, 417
Merck Index 15, 5771
Solubility Information Solubility in water: soluble.
Formula Weight 134.09
Percent Purity 98%
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Malic acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.