Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Lactose Monohydrate (Powder/NF), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
Specifications
| Melting Point | 219°C |
| Color | White |
| Physical Form | Solid |
| Quantity | 500 g |
| Formula Weight | 360.32 |
| Grade | NF |
| Identification | Pass Test |
| Loss on Drying | 0.5% max. |
| Packaging | Poly Bottle |
| Appearance | Pass Test |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction