Learn More
Pfanstiehl Inc D-Glucose (Dextrose), Anhydrous, High Purity, Low Endotoxin, Low Metals, USP, EP, BP, JP, CHP, 1KG, GMP, Multi-compendial, Injectable Excipient Grade (CAS 50-99-7)
Supplier:  Pfanstiehl Inc G1391KG
Encompass
Pfanstiehl Dextrose High Purity, Low Endotoxin, Low Metals is a multi-compendial carbohydrate that is utilized as chemically defined component to optimize protein production. Pfanstiehl Dextrose is specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Dextrose meets parenteral (injectable) excipient grade standards, making it ideal as a bioprocessing feed. Pfanstiehl Dextrose is available as USP (United States Pharmacopoeia) EP (European Pharmacopoeia), and BP (British Pharmacopeia) . Compendial, U.S.P. , cGMP , pharmaceutical grade .
Catalog No. NC3315804