Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Cysteine, 99%

Catalog No. AC254300010
Click to view available options
Quantity:
1 g

Catalog No. AC254300010

Supplier: Thermo Scientific Chemicals 254300010

Only null left
Add to Cart

CAS: 921-01-7 | C3H7NO2S | 121.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 921-01-7
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.15
MDL Number MFCD00066461
InChI Key XUJNEKJLAYXESH-RUOZJJLTNA-N
Synonym d-cysteine, d-cystein, 2s-2-amino-3-sulfanylpropanoic acid, d-zystein, cysteine, d, s-2-amino-3-mercaptopropanoic acid, d-amino-3-mercaptopropionic acid, d-cys, h-d-cys-oh, cysteine
PubChem CID 92851
ChEBI CHEBI:16375
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid
SMILES N[C@H](CS)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula HSCH2CH(NH2)CO2H
Specific Rotation -8.0° to -9.5° (20°C, 589nm) (C=5, 2N HCl)
Quantity 1 g
Formula Weight 121.16
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material D-Cysteine
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.