Learn More
Aat Bioquest Cy7-streptavidin conjugate (1 mg)
SDP
Supplier:  Aat Bioquest 16914
Encompass_Preferred
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins protein motifs nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin. This catalog is for 1 mg.
Catalog No. 50-224-9623