Learn More
Ibidi USA Culture-Insert 2 Well in µ-Dish 35mm, High, ibiTreat - Tissue Culture Treated Polymer Coverslip
Supplier: Ibidi USA 81176
The ibidi Culture-Insert family is developed for wound healing, co-cultivation, and migration assays of adherent cells. The Culture-Insert 2 Well consists of two wells, separated by a 500 µm wall. A proprietary biocompatible adhesive is applied to the underside of the Culture-Insert which allows it to adhere to any flat, clean, and dry surface. A cell suspension can be placed in the wells allowing the adherent cells to grow in the designated areas only. After cell cultivation, the Culture-Insert can be removed by using sterile tweezers. The removal creates a clean, cell-free, and well-defined 500 µm gap. Live-cell imaging can be used to analyze the gap closure. The Culture-Insert 2 Well comes ready-to-use in our µ-Dish 35 mm, High, ibiTreat dishes. sterile, 30 pcs./box.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.