Learn More
Medchemexpress LLC HY-N1405 1mg Medchemexpress, Cucurbitacin I CAS:2222-07-3 Purity:>98%
Supplier: Medchemexpress LLC HYN14051MG
Medchemexpress, HY-N1405 1mg Cucurbitacin I CAS:2222-07-3 Cucurbitacin I is a natural selective inhibitor of JAK2/STAT3 , with potent anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.