Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cresyl Violet acetate

Catalog No. AAJ6431809
Click to view available options
Quantity:
10 g

Catalog No. AAJ6431809

Supplier: Thermo Scientific Chemicals J6431809

Only null left
Add to Cart

CAS: 10510-54-0 | C18H15N3O3 | 321.34 g/mol

Cresyl Violet acetate is used as stain for nerve cells and tissue spinal cord section, brain paraffin, cells and granules. It is also useful to measure pyruvate dehydrogenase complex activity when its acetylation is coupled with acetyl-CoA production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cresyl Violet acetate is used as stain for nerve cells and tissue spinal cord section, brain paraffin, cells and granules. It is also useful to measure pyruvate dehydrogenase complex activity when its acetylation is coupled with ace
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10510-54-0
Molecular Formula C18H15N3O3
Molecular Weight (g/mol) 321.34
MDL Number MFCD00013151,MFCD00013151
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
Synonym 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt; Cresyl echt violet
PubChem CID 44134641
IUPAC Name acetic acid;5-iminobenzo[a]phenoxazin-9-amine
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3

Specifications

Melting Point 140°C to 143°C
Solubility Information Soluble in water and ethanol. Insoluble in acetone.
Formula Weight 321.33
Color Black to Green
Quantity 10 g
Physical Form Powder
Chemical Name or Material Cresyl Violet acetate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-043-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.