Learn More
Teco Diagnostics CREATININE KIT
Supplier: Teco Diagnostics C515480
Creatinine reagent is used for the quantitative determination of creatinine in human serum. 2x120mL Vials of Creatinine Picric Acid, 2x120mL Vials of Creatinine (D) Buffer, 1x10mL Vial of Creatining (D) Standard
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.