Learn More
AllCells LLC Cord Blood- Cryopreserved- CD34+ Stem/Progenitor Cells- Positively Selected- CS10- Mixed- 1M
Supplier:  AllCells LLC CB CR CD34 MIX PS CS10 1M
Encompass
Cord blood from a two or more donors is used to positively select CD34+ cells using an immunomagnetic labeling system. Stem/Progenitor cells have the potential to self-renew and differentiate into all the mature blood cell lineages. The cells are cryopreserved with CS10 in a cryovial. Stem cell purity is determined post thaw using a FACS assay. Cell count and viability are determined using AO/PI. Cells are collected from healthy donors following IRB protocols. Umbilical Cord Blood is a rich source of primitive CD34+ hematopoietic stem and progenitor cells reported to be more naive with a higher proliferative- engraftment and multipotent potential than those isolated from adult bone marrow. Because of these attributes- cord blood can be a great tool for applications where bypassing HLA mismatch is required. AllCells offers cryopreserved cord blood CD34+ from both single cord or multiple/mixed cord donor samples.
Catalog No. NC2742200
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.