Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX7A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549922
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PA549922 100 μL
1 options

Catalog No. PA549922

Supplier: Invitrogen™ PA549922

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total COX7A3 / COX7A2P2 protein.

This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX7A2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COX7A2
Gene Accession No. O60397, P14406, P35171, P48771
Gene Alias Cox7a2; Cox7a3; COX7AL; COX7AL1; COX7AL2; COXVIIAL; COXVIIa-L; cytochrome c oxidase polypeptide 7A2, mitochondrial; cytochrome c oxidase polypeptide VIIa-liver/heart; cytochrome c oxidase subunit 7A2; cytochrome c oxidase subunit 7A2, mitochondrial; cytochrome c oxidase subunit VIIa 2; cytochrome c oxidase subunit VIIa 3; cytochrome c oxidase subunit VIIa polypeptide 2; cytochrome c oxidase subunit VIIa polypeptide 2 (liver); cytochrome c oxidase subunit VIIaL; cytochrome c oxidase subunit VIIa-L; Cytochrome c oxidase subunit VIIa-liver/heart; cytochrome c oxidase, subunit 7a 3; cytochrome c oxidase, subunit VIIa 2; hepatic cytochrome-c oxidase chain VIIa; VIIAL
Gene Symbols COX7A2, COX7A2P2
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human COX7S/A2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12866, 1347, 1348, 29507
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.