Learn More
Covidien In-Room™ Sharps Disposal System SharpStar™ Containers
Catalog No. 22557177
Click to view available options
Capacity (English):
5 qt.
12 qt.
Color:
Clear
Translucent Red
This item is not returnable. View return policy
Catalog No. 22-557-177
Supplier: Covidien 8507SA
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Engineered with controls to prevent overfilling and to limit accidental or intentional access to the container contents
  • Forced horizontal drop maximizes container capacity
  • Anti-kickback feature prevents sharps from exiting the container
  • “FULL” sign signifies that container has reached capacity
  • Locking tabs on top of lid keeps hands away from contents during final closure
  • Molded handles allow safe and secure transport of container
  • Nestable containers save inventory space
Specifications
Capacity (Metric) 4.7 L
Closure Type Counter-Balanced Lid
Capacity (English) 5 qt.
Color Translucent Red
Model Sharpstar
Quantity 20/Cs.
Stackable Yes
Autoclavable Autoclavable
Material Polypropylene
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.