Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate Solution, ASTM A380/A967, Reagents

Catalog No. 01335566
Click to view available options
Quantity:
500 mL
4 L

Catalog No. 01-335-566

Supplier: Reagents Holdings Llc CC256000500A

Only null left
Add to Cart
Add to Cart

Copper Sulfate Solution, ASTM A380/A967, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Boiling Point 100°C
Chemical Name or Material Copper Sulfate Solution, ASTM A380/A967
CAS 7732-18-5, 7758-99-8
Color Blue
pH <1
Physical Form Liquid
Quantity 500 mL
Molecular Formula CuSO4/H2O/H2SO4
Synonym Copper(II) Sulfate
Percent Purity 1.6%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.01
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.