Learn More
Copper Nickel wire, 0.5mm (0.02in) dia
Description
Copper Nickel wire is employed for sea water condenser systems and in desalination plants, as well as pipe work in chemical plants. It is being increasingly employed for brake and hydraulic suspension systems and cooling systems in cars and commercial vehicles. It is commonly used for the manufacture of coins, medals and other semi valuable applications. It is also used in cooling circuits, ammunition, sea water corrosion-resistant assemblies and condenser tubes. The good resistance to biofouling and sea water corrosion resistance of the copper nickel alloys have lead to its use in applications such as cladding for ships' hulls, legs of oil rig platforms and sea water intake screens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Copper Nickel wire |
| Name Note | 0.5mm (0.02 in.) dia. |
| Avg. Mol. Wt. or Mol. Wt. Range | Cu:Ni 55:45 wt% |
| Quantity | 25 m |
| Weight | ≈1.75g/m |
| Form | Wire |
| MDL Number | MFCD00801098 |
| Solubility Information | Insoluble in water. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.