Learn More
Hardy Diagnostics™ Copan Mini-Tip Rayon Swab with aluminum Shaft
Shop All Hardy Diagnostics Products
Click to view available options
Tip Type:
Minitip
Description
- Individually packed
- Suitable for Nasopharyngeal or Oropharyngeal
- Shaft has no break-point – must cut wire manually to fit into transport medium
Specifications
Specifications
| Tip Type | Minitip |
| Handle Material | Aluminum |
| Tip Material | Rayon |
| Sterility | Sterile |
| Packaging | Plastic Round Bottom Tube |
| No. per Case | 1000 |
| Cap Color | Orange |
| Breakpoint | No |
| Product Type | Swab |