Learn More
Research Products International Corp Cool Cube Benchtop Cooler, Reversible, Holds 36 1.5 - 2.0 ml or 12 1.5 - 2.0 ml and PCR Plate
SDP
Supplier:  Research Products International Corp 260105
Encompass
The CoolCube provides convenient storage of heat sensitive samples on a lab bench and maintains temperature at 0°C for up to four hours. Constructed of a high strength polymer that has excellent chemical resistance and filled with a gel refrigerant. Simply place the rack in a -20°C freezer overnight and it is ready to use. Samples remain cool without the use of messy wet ice.
One side holds 36 x 1.5-2.0 ml micro-tubes. Opposite side accommodates 12 x 1.5-2.0 ml micro-tubes and one PCR plate.
Catalog No. 50-550-329
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.