Description
- Room temperature storage
- Enhanced sample preparation procedure
- Results in 6 minutes
- Sample from culture (identified as Staphylococcus aureus)
- Built-in procedural controls on every test strip
Specifications
Specifications
| Clia Complexity | Moderate |
| Detectable Analytes | PBP2a Protein |
| Includes | Test strips, reagent 1, reagent 2, assay tubes, and test racks |
| Quantity | 25 Tests |
| Test Time | 6 min. |