Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cisapride
GREENER_CHOICE

Catalog No. 169510
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 16-951-0

Supplier: Tocris Bioscience™ 1695/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

5-HT4 agonist; stimulates intestinal ACh release

5-HT4 receptor agonist and gastrokinetic agent. Stimulates intestinal acetylcholine release, possibly via 5-HT4 receptor-dependent and -independent mechanisms, leading to increased intestinal motility.

Chemical Identifiers

CAS 81098-60-4
Molecular Formula C23H29ClFN3O4

Specifications

Quantity 10 mg
Formula Weight 465.95
Purity >99%
Molecular Weight (g/mol) 465.95
Chemical Name or Material Cisapride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.