Learn More
Active Motif Chromatin IP DNA Purification Kit
SDP
Supplier:  Active Motif 58002
Encompass
Active Motif's Chromatin IP DNA Purification Kit allows you to quickly clean up your ChIP DNA samples and get them ready for analysis, without the need for messy, labor intensive and time consuming phenol/chloroform extraction. Our method is optimized for chromatin IP and epigenetics applications and completely compatible with samples generated using our ChIP-IT chromatin immunoprecipitation kits.
Catalog No. NC0116263