Learn More
ATCC CHO-K1; Ovary; Chinese Hamster (Cricetulus griseus)
Supplier: ATCC CCL61
CHO-K1 cell line was derived as a subclone from the parental CHO cell line, which was initiated from a biopsy of an ovary of an adult, female Chinese hamster in 1957. The cell line is used for industrial biotechnology and toxicology research.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.