Learn More
Chembio HIV 1/2 STAT-PAK™ Assay
GSA_VASDP
Catalog No. 23999102
Change view
Click to view available options
Type:
HIV 1/2 STAT-PAK Assay
Catalog No. Type
23-999-102 HIV 1/2 STAT-PAK Assay
1 options
This item is not returnable. View return policy
Catalog No. 23-999-102 Supplier Chembio Supplier No. 6095051
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Easy-to-perform, single-use diagnostic tests for the rapid (15 minutes), visual detection of antibodies to HIV 1 and HIV 2 at point-of-care (POC)
Convenient and Cost Effective
  • Ideally suited for point-of-care testing
  • Minimal sample size required—5μl
  • Room temperature storage, 8° to 30° C (46° to 86° F)
  • Long shelf life
Time and Labor Saving
  • Ready-to-Use reagents (no special preparation of reagents required)
  • Simple procedure
  • Rapid reporting: Results in 15 minutes
  • No special equipment required
  • Easy-to-interpret results
Reliable Results
  • Built-in IgG procedural control
  • 99.7% sensitivity and 99.9% specificity (see product insert for complete performance details)
For Use With:
  • Serum
  • Plasma
  • Fingerstick whole blood
  • Venipuncture whole blood
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Clia Complexity Waived, Non-waived
Type HIV 1/2 STAT-PAK Assay
Includes 20 Individually Pouched Test Devices; 20 Disposable 5 μL Sample Loops;1 HIV Running Buffer (3.5 mL); 20 Copies of Subject Information Notice; 1 Quick Reference Instuctions (for CLIA Waived); 1 Product Insert