Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coastline Global™ CheckUp 10-in-1 Dog and Cat Urine Testing Strips, 50 count
Click to view available options
Detectable Analytes:
Glucose, Bilirubin, Ketone, Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leucocytes
Description
50 Individual diagnostic dog or cat urine CheckUp testing strips for a variety of parameters that correspond with different medical conditions.
- Urobilinogen
- Glucose
- Bilirubin
- Ketone
- Specific Gravity
- Blood
- pH
- Protein
- Nitrite
- Leukocytes
Specifications
Specifications
| CE Marker | Self-declared |
| Clia Complexity | Waived (Urine) |
| Detectable Analytes | Glucose, Bilirubin, Ketone, Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leucocytes |
| Form | Ready-to-Use (RTU) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction