Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CGP 35348

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective, brain penetrant GABAB receptor antagonist (IC50 = 34 μM as measured in rat cortical membranes). Has higher affinity for postsynaptic versus presynaptic receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 225.22 |
| Purity | >98% |
| Molecular Weight (g/mol) | 225.23 |
| Chemical Name or Material | CGP 35348 |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction