Learn More
Santa Cruz Biotechnology CGAS ANTIBODY (C-1) 200UG/ML
Supplier: Santa Cruz Biotechnology SC515802
The presence of foreign DNA in the cytoplasm induces an antiviral host immune response. DNA in the cytoplasm triggers the production of interferons by activating and synthesis of second messenger cyclic guanosine monophosphate-adenosine monophosphate (cyclic GMP-AMP, or cGAMP). cGAS (cyclic GMP-AMP synthase), also known as MB21D1 (Mab-21 domain containing 1), h-cGAS or C6orf150, is a 522 amino acid cytoplasmic nucleotidyltransferase that catalyzes the formation of cyclic GMP-AMP (cGAMP) from ATP and GTP. cGAS is suggested to have antiviral activity by acting as a key cytosolic DNA sensor. cGAS binds to cytosolic DNA, which leads to cGAMP synthesis and activation of TMEM173, thereby trigger type-I interferon production. Expressed in monocytic cell line THP1, cGAS exists as two alternatively spliced isoforms and is encoded by a gene located on human chromosome 6q13.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.