Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cetirizine Impurity Standard, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
:
50 mg
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 83881-52-1 |
| Molecular Formula | C21H25ClN2O3 · 2 HCl |
| Linear Formula | C21H25ClN2O3 · 2 HCl |
| MDL Number | MFCD00941428 |
| Quantity | 50 mg |
| Synonym | Cetirizine dihydrochloride; [2-[4-[(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]acetic acid dihydrochloride |
| Formula Weight | 461.81 |
| Grade | British Pharmacopoeia (BP) Reference Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction