Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cereal Grass Medium
SCI_ED

Catalog No. s25242a Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g

Catalog No. S25242A

Only null left
Add to Cart
Add to Cart

For culturing protozoa (rhizopods, choanoflagellates, ciliates and flagellates).

  • Contains dehydrated cereal grass leaves with natural vitamins A, B, C, and K
  • Recommended by King's lab at The University of California at Berkeley
  • DOT Info: Non-regulated

This product is for educational use - no C of A is available.

Specifications

Form Powder
Media Type Cereal Grass Medium
Product Type Cereal Grass Medium
For Use With (Application) Cultivation of protozoa (rhizopods, choanoflagellates, ciliates and flagellates)
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.