Learn More
CEP131/AZ1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00363
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee and Northern white-cheeked gibbon
CEP131 (Centrosomal Protein 131) is a Protein Coding gene. Diseases associated with CEP131 includeAnterior Spinal Artery Syndrome. Among its related pathways areOrganelle biogenesis and mainteceandCell Cycle, Mitotic. Gene Ontology (GO) annotations related to this gene includeprotein homodimerization activity.Specifications
| CEP131/AZ1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9UPN4 | |
| CEP131 | |
| Between 1033 and 1083 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| CEP131 | |
| 5-azacytidine induced 1, 5-azacytidine-induced protein 1, AZ1, AZI1, centrosomal protein 131 kDa, centrosomal protein 131kDa, KIAA1118, pre-acrosome localization protein 1, ZA1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22994 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.