Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega CellTiter-Glo™ 2.0 Assay

Catalog No. PRG9243 Shop All Promega Corporation Products
Click to view available options
Quantity:
10 mL
100 mL
500 mL

Catalog No. PRG9243

Supplier: Promega G9243

Only null left
Add to Cart
Add to Cart

The CellTiter-Glo™ 2.0 Assay provides a homogeneous method of determining the number of viable cells in culture by quantitating the amount of ATP present, which indicates the presence of metabolically active cells.

  • A Luminescent Cell Viability Assay for Fast, Easy, Everyday Use
  • Reduced preparation time
  • Convenient repeat use over multiple days
  • Same great performance as classic CellTiter-Glo(R) Assay

Specifications

Quantity 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.