Learn More
GLPBIO TECHNOLOGY INC Cell Counting Kit-8 (CCK-8), 100 mL
Supplier: GLPBIO TECHNOLOGY INC GK10001100ML
Cell Counting Kit-8 (CCK-8) is a ready-to-use one-bottle solution which offers a simple, rapid, reliable and sensitive measurement of cell viability and cytotoxicity of various chemicals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.