Learn More
Medchemexpress LLC HY-139450 5mg , CDK4/6-IN-6 CAS:2380321-51-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1394505MG
Encompass_Preferred
HY-139450 5mg CDK4/6-IN-6 CAS: 2380321-51-5 CDK4/6-IN-6 (example A94) is a potent CDK4/CDK6 inhibitor with a Ki of 0.6 nM and 13.9 nM for CDK4/Cyclin D1 and CDK6/Cyclin D3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1822
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.