Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK11 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7143980
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1368 10 μL
50-157-1367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1368 Supplier Bethyl Laboratories, Inc Supplier No. IHC00035T
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda and Orangutan

The PITSLRE beta1 protein, a distantly related member of the Cdk family of protein kinases, induces apoptosis after low levels of ectopic expression. Apoptosis, or programmed cell death, is similarly induced by ectopic expression of an amino terminal deletion mutant retaining the catalytic and carboxyterminal domains of PITSLRE beta1, but not by other mutants lacking Histone H1 kinase activity or by other Cdk family members. The terminology for the ten isoforms of the PITSLRE subfamily of proteins is based on the conserved PSTAIRE box region of Cdc2 p34. Depending on which of the PITSLRE genes produce the protein, the cDNA and protein are designated alpha, beta or gamma (i.e., PITSLRE A gene, alpha; PITSLRE B gene, beta and PITSLRE C gene, gamma). Some of the isoforms such as PITSLRE alpha1 (T cells) and PITSLRE beta1 (B cells and brain), are expressed in specific cell types, while others are expressed ubiquitously.

Specifications

Antigen CDK11
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CDK11A
Gene Accession No. Q9UQ88
Gene Alias CDC2L2, CDC2L3, CDK11, CDK11 p110, CDK11 p46, CDK11 p58, CDK11A, CDK8 like cyclin dependent kinase, Cell division cycle 2 like 2 (PITSLRE proteins), Cell division cycle 2 like protein kinase 2, Cell division cycle 2-like protein kinase 2, Cell division protein kinase 11A, Cyclin dependent kinase (CDC2 like) 11, Cyclin-dependent kinase 11A, Death preventing kinase, Galactosyltransferase associated protein kinase p58/GTA, Galactosyltransferase-associated protein kinase p58/GTA
Gene Symbols CDK11A
Host Species Rabbit
Immunogen Between 730 and C-term
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728642
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.