Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561072
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA561072 100 μL
1 options

Catalog No. PIPA561072

Supplier: Invitrogen™ PA561072

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEFIEMKSNL KHLEVLVAQQ SQEFQQLCEQ LGQLNVPSVL AELKRLISVP PVKDSASQTS PPLAQSLNLT RQEKYTSEKP VLWQAQALPA AWNPGMGSLQ Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 61%.

Required for DNA double-strand breaks (DSBs) formation in unsynapsed regions during meiotic recombination. Probably acts by forming a complex with MEI4 and REC114, which activates DSBs formation in unsynapsed regions, an essential step to ensure completion of synapsis. Not required for HORMAD1 functions in pairing-independent synaptonemal complex formation, ATR recruitment to unsynapsed axes, meiotic silencing of unsynapsed chromatin (MSUC) or meiotic surveillance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC36
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC36
Gene Accession No. Q8IYA8
Gene Alias Cancer/testis antigen 74; Ccdc36; coiled-coil domain containing 36; coiled-coil domain-containing protein 36; CT74; IHO1; Interactor of HORMAD1 protein 1
Gene Symbols CCDC36
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC36. Recombinant protein control fragment (Product #RP-100228).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.