Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cav beta Ca+2 Channel Monoclonal Antibody (S8b-1), QED Bioscience™
SDP

Catalog No. 502277437
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7437 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7437 Supplier Leinco Technologies Inc Supplier No. 11524100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cav beta Ca+2 Channel Monoclonal antibody specifically detects Cav beta Ca+2 Channel in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Cav beta Ca+2 Channel
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone S8b-1
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Cacnb2
Gene Accession No. Q08289, Q8CC27, Q8VGC3
Gene Alias AW060387; brain calcium channel beta-subunit CaB2c (splice variant of the gene CaB2); CAB1; CAB2; Cacnb2; CACNLB1; Cacnlb2; Calcium channel voltage-dependent subunit beta 2; calcium channel, voltage-dependent, beta 2 subunit; calcium voltage-gated channel auxiliary subunit beta 2; cardiac calcium channel beta subunit; cardiac calcium channel beta-subunit CaB2a (splice variant of the gene CaB2; cardiac calcium channel beta-subunit CaB2b (splice variant of the gene CaB2); CAVB2; Cavbeta2; Cchb2; CCHLB1; lambert-Eaton myasthenic syndrome antigen B; L-type calcium channel beta subunit; L-type voltage-dependent calcium channel beta-2 subunit; MGC138502; MGC138504; MGC41896; myasthenic (Lambert-Eaton) syndrome antigen B; MYSB; voltage-dependent calcium channel beta2A subunit; voltage-dependent calcium channel beta2B subunit; voltage-dependent L-type calcium channel subunit beta-2; voltage-gated calcium channel beta2 subunit transcript variant X13
Host Species Mouse
Immunogen Synthetic peptide corresponding to aa 189-205 of rat Cav beta 2 (RSPKPSANSVTSPHSKE) (accession no. NP_446303).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116600, 12296, 783
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.