Learn More
Biomatik Corporation GALNT15 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09506
Encompass_Preferred
The GALNT15 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Polypeptide N-acetylgalactosaminyltransferase 15 protein (35-335AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Polypeptide N-acetylgalactosaminyltransferase 15, Polypeptide GalNAc transferase-like protein 2, GalNAc-T-like protein 2, pp-GaNTase-like protein 2, Polypeptide N-acetylgalactosaminyltransferase-like protein 2, Protein-UDP acetylgalactosaminyltransferase-like protein 2, polypeptide N-acetylgalactosaminyltransferase-like protein 2, GALNT15, GALNTL2, UNQ770/PRO1564. UniProt: Q8N3T1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9088