Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Castor Oil, USP, Fisher Chemical

Catalog No. o464 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L

Catalog No. O46-4

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-79-4
Molecular Formula C57H104O9
Molecular Weight (g/mol) 933.45
MDL Number MFCD00130746
InChI Key ZEMPKEQAKRGZGQ-AAKVHIHISA-N
Synonym castor oil, ricinus oil, olio di ricino, venelex, xenaderm, optase, trypsin complex, unii-d5340y2i9g, castor oil usp:jan, 1-o,2-o,3-o-tris z-12-hydroxy-9-octadecenoyl glycerol
PubChem CID 14030006
IUPAC Name 2,3-bis[[(Z)-12-hydroxyoctadec-9-enoyl]oxy]propyl (Z)-12-hydroxyoctadec-9-enoate
SMILES CCCCCCC(CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(CCCCCC)O)OC(=O)CCCCCCCC=CCC(CCCCCC)O)O

Specifications

Acidity NMT 2.0
Melting Point -10°C
Color Yellow
Density 0.96g/mL
Boiling Point 229°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.957 to 0.961 (USP)
Residual Solvents Meets Requirements (USP)
Grade USP
Physical Form Liquid
Chemical Name or Material Castor Oil,
  • Unsaponifiable Matter: NMT 0.8%
  • Procedures Acid Value: NMT 2.0
  • Procedures Peroxides Values: NMT 10.0
  • Water determination: NMT 0.3%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.