Learn More
Description
- For a class of 30 students
- A large species of Daphnia
- Commonly used in aquatic effluent toxicity testing
- An obligate algal feeder
- Cultured using powdered Spirulina as a food source
Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.
Specifications
Specifications
| For Use With (Application) | Commonly used in aquatic effluent toxicity testing, obligate algal feeder. |
| Storage Requirements | 21°C |
| Quantity | Ea. |
| Scientific Name | Daphnia magna |
| Includes | Adults, juveniles, neonates |
| Sufficient For | Class of 30 students |
| Product Type | Daphnia Magna |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

