Learn More
Foxx Life Sciences EZclear 10L (2.5 Gal) BPA-Free PETG Space Saving Carboy with 83mm (83B) VersaCap and 13L Max Capacity with Spigot Large Pinched Handles
Supplier: Foxx Life Sciences 1572211OEM
The EZClear Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows the user to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle as well as large, easy to read, metric graduation marks which are certified to ±5% accuracy.PETG carboys are made from a USP Class VI, FDA Grade, BPA-Free materials.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.