Learn More
Pfanstiehl Inc D-Mannitol, (Plant Derived) High Purity, Low Endotoxin, Low Metals USP, EP, JP, ChP (M-109-7) - 1Kg pack GMP, Multi-compendial CAS: 69-65-8 Injectable Excipient Grade
Supplier:  Pfanstiehl Inc M10971KG
Encompass
Pfanstiehl D-Mannitol High Purity, Low Endotoxin, Low Metals is a multi-compendial sugar alcohol derived from mannose by reduction. Pfanstiehl D-Mannitol has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Mannitol meets parenteral (injectable) excipient grade standards, making it ideal in pharmaceutical formulation. Mannitol's physical properties make it an ideal bulking agent for tableting, while its unique chemical stability allows it to support a wide range of pharmaceutical formulations, including liquid preparations. It is commonly used to stabilize proteins throughout formulation, freeze-drying, and fill/finish processes, ensuring product integrity and efficacy. Pfanstiehl Mannitol is available as USP (United States Pharmacopoeia), EP (European Pharmacopoeia), JP (Japanese Pharmacopoeia), and ChP (Chinese Pharmacopoeia).
Catalog No. 50-165-6644