Learn More
Pfanstiehl Inc Sucrose, USP-NF, EP, BP, JP, ChP High Purity, Low Endotoxin, Low Metals (Beet Derived) (S-124-2-MC) 1kg pack GMP, Multi-Compendial CAS: 57-50-1 Injectable Excipient Grade
Supplier:  Pfanstiehl Inc S1242MC1KG
Encompass
Pfanstiehl Sucrose (Beet Derived) - High Purity, Low Endotoxin, Low Metals is a multi-compendial stabilizer of proteins, lipids, and carbohydrates in biologic therapeutics. Used throughout formulation and freeze/thaw cycles, it ensures stability and efficacy in biopharmaceutical applications. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Sucrose meets parenteral (injectable) excipient grade standards, making it ideal for biologic and vaccine formulations. It is also widely used in cryopreservation media to protect cell membranes and enhance cell recovery for bioproduction and cell therapy. Pfanstiehl Brand Sucrose is multi-compendial and available as USP-NF (National Formulary), EP (Ph. Eur.) (European Pharmacopoeia), JP (Japanese Pharmacopoeia), and ChP (Chinese Pharmacopoeia) Compendial Sucrose, GMP Sucrose, cGMP sucrose, sucrose excipient, injectable sucrose, pharmaceutical sucrose, U.S.P.
Catalog No. 50-165-6664