Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capping Reagent A, Novabiochem™, MilliporeSigma™

Catalog No. MBI02214000 Shop All MilliporeSigma Products
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Catalog No. MBI02214000

Supplier: MilliporeSigma BI02214000

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetyl oxide, acetic acid, anhydride, acetic oxide, acetyl anhydride, anhydride acetique, ethanoic anhydride, acetyl ether, acetic acid anhydride
PubChem CID 7918
ChEBI CHEBI:36610

Specifications

Flash Point 4°C
Packaging Glass Bottle
Quantity 4 L
Percent Purity 9.0-11.0%
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.