Learn More
CAMSAP1L1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | CAMSAP1L1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-1930
|
Bethyl Laboratories, Inc
A302162AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
CAMSAP2 gene ontology annotations related to this gene include neuron projection development.Specifications
| CAMSAP1L1 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| calmodulin regulated spectrin-associated protein 1-like 1, calmodulin regulated spectrin-associated protein family, member 2, calmodulin-regulated spectrin-associated protein 1-like protein 1, CAMSAP1L1, KIAA1078 | |
| CAMSAP2 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q08AD1 | |
| 23271 | |
| Between 1439 and 1489 | |
| Primary | |
| 4° C | |
| CAMSAP2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.