Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
calcium homeostasis modulator 2 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | calcium homeostasis modulator 2 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
calcium homeostasis modulator 2 Polyclonal specifically detects calcium homeostasis modulator 2 in Rat samples. It is validated for Western Blot.Specifications
| calcium homeostasis modulator 2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| calcium homeostasis modulator 2, calcium homeostasis modulator protein 2, FAM26Bfamily with sequence similarity 26, member B, Protein FAM26B | |
| The immunogen is a synthetic peptide directed towards the C terminal region of rat calcium homeostasis modulator 2 (NP_001008307). Peptide sequence EELVTKFPVEGTQPRPQWNAITGVYLYRENQGLPLYSRLHKWAQGLTGNG | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 51063 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title