Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Glycerophosphate Hydrate 95.0+%, TCI America™
SDP

Catalog No. G009525G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Catalog No. G009525G

Supplier: TCI America G009525G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27214-00-2
Molecular Formula C3H7CaO6P
Molecular Weight (g/mol) 210.135
MDL Number MFCD00042643
InChI Key IWIRHXNCFWGFJE-UHFFFAOYSA-L
Synonym Glycerophosphoric Acid Calcium Salt
PubChem CID 120096
IUPAC Name calcium;2,3-dihydroxypropyl phosphate
SMILES C(C(COP(=O)([O-])[O-])O)O.[Ca+2]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 210.13
Percent Purity ≥95.0% (T)
Chemical Name or Material Calcium Glycerophosphate Hydrate
EINECSNumber (2)-1905
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.