Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BSA, 10% Aqueous Solution, Nuclease-Free

Catalog No. 12661525ML Shop All MilliporeSigma Products
Click to view available options
Quantity:
25 mL

Catalog No. 12-661-525ML

Supplier: MilliporeSigma™ 12661525ML

Only null left
Add to Cart
Add to Cart

Reduces DNases RNases to low levels.

  • Applications: Antibody dilution, DNA foot printing, gel shift assay, ELISA, enzyme assay, enzyme stabilization, immunoblotting, immunofluorescence, PCR, restriction enzyme reactions, RIA, probe-based diagnostics, radioactive quenching, receptor binding studies
  • Contaminants
  • Ash: ≤2.0%
  • Fatty Acids: ≤0.02%
  • Nuclease: Non-Detected
  • Protease Activity: Non-Detected
  • Heavy Metal: ≤10ppm
  • CAS: 9048-46-8

Recommended Storage

2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Description BSA
Formulation 10% aqueous solution, 0.22μm-filtered, pH 6.8-7.2.
Quantity 25 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.