Learn More
Chemglass Life Sciences Cable Tie, Zip Tie, 4" Long, Nylon 6.6, Natural, Autoclavable
Supplier: Chemglass Life Sciences CLS1399T100
These tie wraps are injection-molded of 6/6 nylon and won't corrode or stretch. Durable locking mechanism holds securely. Color: Natural. Strength: 18lb. Length: 4” (100mm). Width: 1/8” (3mm). Max Bundle/Wrap Diameter: 1” (25mm). Temperature: Long term: -40 to 85 degrees C. Autoclavable: YES..Please Note: Ties are intended for single use and can only be autoclaved once.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.