Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C7 - C40 Saturated Alkanes, Standard Certified Reference Material, 1,000 μg/mL Each Component in Hexane, MilliporeSigma™ Supelco™

Catalog No. 111017207 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL

Catalog No. 11-101-7207

Supplier: MilliporeSigma Supelco 49452U

Only null left
Add to Cart
Add to Cart

Quantitative standard suitable for use as a retention index marker probe.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Concentration 1000 μg/mL each component in hexane
Quantity 1 mL
Grade Certified Reference Material
Concentration or Composition (by Analyte or Components) Decane; Docosane; Dodecane; Dotriacontane; Eicosane; Heneicosane; Hentriacontane; Heptacosane; Heptadecane; Heptane; Heptatriacontane; Hexacosane; Hexadecane; Hexatriacontane; Nonacosane; Nonadecane; Nonane; Nonatriacontane; Octacosane; Octadecane; Octane; Octatriacontane; Pentacosane; Pentadecane; Pentatriacontane; Tetracontane; Tetracosane; Tetradecane; Tetratriacontane; Triacontane; Tricosane; Tridecane; Tritriacontane; Undecane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.