Learn More
Medchemexpress LLC HY-120934 5mg Medchemexpress, C25-140 CAS:1358099-18-9 Purity:>98%
Supplier: Medchemexpress LLC HY1209345MG
Medchemexpress, HY-120934 5mg C25-140 CAS:1358099-18-9 C25-140, a first-in-class, fairly selective TRAF6-Ubc13 inhibitor, directly binds to TRAF6, and blocks the interaction of TRAF6 with Ubc13. C25-140 lowers TRAF6 activity, reduces NF-κB activation, and combats autoimmunity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.