Learn More
Abcam Human Complement C1q ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1702461X96TESTS
Encompass_Preferred
Human Complement C1q ELISA Kit ab170246 is used to quantify Human Complement C1q with a sensitivity of 64 pg/ml. Sandwich ELISA Kit - Validated in a range of biological samples including CSF
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1222930
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.